Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ttr-19
Entrez Gene ID
182540
Gene Name
TransThyretin-Related family domain
Gene Aliases
CELE_C12D8.4
Chromosome Location
Chr.V: 10226828 - 10227433 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073229.4 NP_505630.1 1 - 2 174 87

View Full Product Details