Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-2
Entrez Gene ID
179450
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_AC3.8
Chromosome Location
Chr.V: 10398717 - 10401307 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073271.3 NP_505672.2 4 - 5 469 76

View Full Product Details