Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
vps-54
Entrez Gene ID
179473
Gene Name
protein-coding
Gene Aliases
CELE_T21C9.2
Chromosome Location
Chr.V: 10566365 - 10576419 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269297.2 NP_001256226.1 8 - 9 3093 100
NM_001269298.3 NP_001256227.1 3 - 4 2153 100

View Full Product Details