Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-34
Entrez Gene ID
185120
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_F29F11.2
Chromosome Location
Chr.V: 10654415 - 10656978 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073327.6 NP_505728.2 6 - 7 1347 94

View Full Product Details