Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ugt-35
Entrez Gene ID
3565443
Gene Name
UDP-GlucuronosylTransferase
Gene Aliases
CELE_C32C4.7
Chromosome Location
Chr.V: 10651995 - 10654265 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028523.1 NP_001023694.1 3 - 4 718 116

View Full Product Details