Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
secs-1
Entrez Gene ID
179498
Gene Name
protein-coding
Gene Aliases
CELE_D1054.13
Chromosome Location
Chr.V: 10802336 - 10804474 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001136415.3 NP_001129887.1 2 - 3 222 102
NM_073360.4 NP_505761.1 1 - 2 96 102

View Full Product Details