Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-31
Entrez Gene ID
186493
Gene Name
protein-coding
Gene Aliases
CELE_F58B4.1
Chromosome Location
Chr.V: 10918857 - 10921847 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028823.6 NP_001023994.1 12 - 13 1689 142

View Full Product Details