Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-41
Entrez Gene ID
188592
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T19C4.3
Chromosome Location
Chr.V: 11160452 - 11161891 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073436.1 NP_505837.1 3 - 4 508 158

View Full Product Details