Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-40
Entrez Gene ID
188593
Gene Name
Serpentine receptor class gamma
Gene Aliases
CELE_T19C4.4
Chromosome Location
Chr.V: 11163206 - 11164575 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073437.4 NP_505838.2 4 - 5 824 63

View Full Product Details