Assay Design

The assay probe spans an exon junction.

Gene Transcripts

Gene Symbol
mxl-1
Entrez Gene ID
179557
Gene Name
BHLH domain-containing protein
Gene Aliases
CELE_T19B10.11
Chromosome Location
Chr.V: 11244482 - 11245930 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073455.6 NP_505856.1 2 - 3 476 103
GenBank mRNA U82967.1 - 1 - 2 322 103

View Full Product Details