Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-17
Entrez Gene ID
186923
Gene Name
Zinc metalloproteinase nas-17
Gene Aliases
CELE_K03B8.2
Chromosome Location
Chr.V: 11398916 - 11400563 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073491.2 NP_505892.1 4 - 5 1158 109

View Full Product Details