Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
nas-19
Entrez Gene ID
186926
Gene Name
Zinc metalloproteinase nas-19
Gene Aliases
CELE_K03B8.5
Chromosome Location
Chr.V: 11401091 - 11402710 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073492.4 NP_505893.2 2 - 3 1065 102

View Full Product Details