Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
tre-3
Entrez Gene ID
179604
Gene Name
protein-coding
Gene Aliases
CELE_W05E10.4
Chromosome Location
Chr.V: 11709769 - 11715903 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269432.3 NP_001256361.1 7 - 8 1409 151

View Full Product Details