Assay Design
The assay probe spans an exon junction. This assay will detect gDNA.
Gene Transcripts
- Gene Symbol
- kup-1
- Entrez Gene ID
-
179651
- Gene Name
- Protein kup-1
- Gene Aliases
-
CELE_F10C2.2
- Chromosome Location
-
Chr.V: 12014433 - 12040873 on Build WBcel235
Interrogated Sequence |
Translated Protein |
Exon Boundary |
Assay Location |
Amplicon Length |
RefSeq |
NM_001383418.1 |
|
10 - 11 |
1628 |
63 |
- Gene Symbol
- pkc-1
- Entrez Gene ID
-
179649
- Gene Name
- protein-coding
- Gene Aliases
-
CELE_F57F5.5
- Chromosome Location
-
Chr.V: 12014383 - 12032165 on Build WBcel235
View Full Product Details