Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clh-6
Entrez Gene ID
179655
Gene Name
Chloride channel protein
Gene Aliases
CELE_R07B7.1
Chromosome Location
Chr.V: 12060233 - 12063368 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073621.5 NP_506022.1 1 - 2 37 61
GenBank mRNA AF319616.1 - 1 - 2 59 61
AF328908.1 - 1 - 2 68 61

View Full Product Details