Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clh-6
Entrez Gene ID
179655
Gene Name
Chloride channel protein
Gene Aliases
CELE_R07B7.1
Chromosome Location
Chr.V: 12060233 - 12063368 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073621.5 NP_506022.1 9 - 10 2294 89
GenBank mRNA AF319616.1 - 9 - 10 2316 89
AF328908.1 - 9 - 10 2325 89

View Full Product Details