Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cdr-6
Entrez Gene ID
179703
Gene Name
CaDmium Responsive
Gene Aliases
CELE_K01D12.12
Chromosome Location
Chr.V: 12411939 - 12413701 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073712.6 NP_506113.1 5 - 6 690 133
GenBank mRNA AY728060.1 - 5 - 6 710 133

View Full Product Details