Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sel-1
Entrez Gene ID
179720
Gene Name
Suppressor/Enhancer of Lin-12
Gene Aliases
CELE_F45D3.5
Chromosome Location
Chr.V: 12559772 - 12563462 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073743.7 NP_506144.1 3 - 4 395 71
GenBank mRNA U50829.1 - 2 - 3 297 71

View Full Product Details