Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-8
Entrez Gene ID
179731
Gene Name
Guanine nucleotide-binding protein alpha-8 subunit
Gene Aliases
CELE_F56H9.3
Chromosome Location
Chr.V: 12638666 - 12647432 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073759.3 NP_506160.1 9 - 10 930 117
GenBank mRNA AY008131.1 - 5 - 6 499 117

View Full Product Details