Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-9
Entrez Gene ID
191659
Gene Name
Guanine nucleotide-binding protein alpha-9 subunit
Gene Aliases
CELE_F56H9.4
Chromosome Location
Chr.V: 12645477 - 12649313 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NR_132533.1 1 - 2 153 116

View Full Product Details