Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gpa-9
Entrez Gene ID
191659
Gene Name
Guanine nucleotide-binding protein alpha-9 subunit
Gene Aliases
CELE_F56H9.4
Chromosome Location
Chr.V: 12645477 - 12649313 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269515.3 NP_001256444.1 2 - 3 225 88
NR_132533.1 8 - 9 1724 88

View Full Product Details