Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
ccch-3
Entrez Gene ID
179733
Gene Name
C3H1-type domain-containing protein
Gene Aliases
CELE_W05B10.2
Chromosome Location
Chr.V: 12658668 - 12661651 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001383424.1 3 - 4 369 123

View Full Product Details