Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-44
Entrez Gene ID
191177
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_ZC455.9
Chromosome Location
Chr.V: 12787269 - 12788551 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073797.1 NP_506198.1 1 - 2 180 118

View Full Product Details