Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
uig-1
Entrez Gene ID
179808
Gene Name
protein-coding
Gene Aliases
CELE_F32F2.1
Chromosome Location
Chr.V: 13273157 - 13295978 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269559.3 NP_001256488.1 6 - 7 1012 141
NM_001269560.3 NP_001256489.1 6 - 7 1054 141
NM_001269563.3 NP_001256492.1 2 - 3 463 141
NM_001380797.1 NP_001367249.1 6 - 7 994 141
NM_001380798.1 NP_001366647.1 6 - 7 1146 141

View Full Product Details