Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srd-11
Entrez Gene ID
191801
Gene Name
protein-coding
Gene Aliases
CELE_F53F4.9
Chromosome Location
Chr.V: 13617717 - 13619852 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073973.2 NP_506374.1 3 - 4 600 107

View Full Product Details