Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
letm-1
Entrez Gene ID
179854
Gene Name
protein-coding
Gene Aliases
CELE_F58G11.1
Chromosome Location
Chr.V: 13661255 - 13664233 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_073980.6 NP_506381.1 5 - 6 2313 64

View Full Product Details