Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sel-10
Entrez Gene ID
179878
Gene Name
protein-coding
Gene Aliases
CELE_F55B12.3
Chromosome Location
Chr.V: 13818114 - 13823091 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028804.4 NP_001023975.1 5 - 6 475 84
NM_074020.7 NP_506421.1 5 - 6 476 84
NR_003169.1 4 - 5 511 84
GenBank mRNA AF020788.1 - 4 - 5 399 84

View Full Product Details