Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-3
Entrez Gene ID
182290
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_C06B3.2
Chromosome Location
Chr.V: 13899513 - 13902653 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074045.2 NP_506446.2 4 - 5 614 74

View Full Product Details