Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-42
Entrez Gene ID
182296
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_C06B3.11
Chromosome Location
Chr.V: 13921030 - 13922404 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074054.2 NP_506455.2 2 - 3 215 147

View Full Product Details