Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sre-7
Entrez Gene ID
186893
Gene Name
Serpentine Receptor, class E (epsilon)
Gene Aliases
CELE_K02E11.2
Chromosome Location
Chr.V: 14244204 - 14246104 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074121.1 NP_506522.1 2 - 3 278 89

View Full Product Details