Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
bmk-1
Entrez Gene ID
179948
Gene Name
protein-coding
Gene Aliases
CELE_F23B12.8
Chromosome Location
Chr.V: 14453302 - 14456998 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001269657.2 NP_001256586.1 7 - 8 2624 75
NM_001269658.3 NP_001256587.1 9 - 10 2683 75
GenBank mRNA AB024428.2 - 7 - 8 2630 75
AF295093.1 - 7 - 8 2615 75

View Full Product Details