Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srg-47
Entrez Gene ID
183738
Gene Name
Serpentine receptor class gamma-47
Gene Aliases
CELE_C53A5.8
Chromosome Location
Chr.V: 14554980 - 14556287 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074203.2 NP_506604.2 4 - 5 827 115

View Full Product Details