Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
wrt-7
Entrez Gene ID
191504
Gene Name
WaRThog (hedgehog-like family)
Gene Aliases
CELE_ZK1037.10
Chromosome Location
Chr.V: 15334754 - 15337505 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074402.4 NP_506803.2 4 - 5 843 88

View Full Product Details