Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srx-29
Entrez Gene ID
187057
Gene Name
7TM_GPCR_Srx domain-containing protein
Gene Aliases
CELE_K06B4.9
Chromosome Location
Chr.V: 15696785 - 15698199 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074503.2 NP_506904.2 2 - 3 202 110

View Full Product Details