Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-27
Entrez Gene ID
188782
Gene Name
Seven TM Receptor
Gene Aliases
CELE_T23D5.1
Chromosome Location
Chr.V: 15734023 - 15735820 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001322595.1 NP_001309461.1 5 - 6 951 74

View Full Product Details