Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srbc-50
Entrez Gene ID
186205
Gene Name
Serpentine Receptor, class BC (class B-like)
Gene Aliases
CELE_F54B8.8
Chromosome Location
Chr.V: 15823898 - 15824966 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074552.2 NP_506953.2 2 - 3 640 145

View Full Product Details