Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-59
Entrez Gene ID
186771
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_H24D24.1
Chromosome Location
Chr.V: 15949716 - 15951003 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074595.1 NP_506996.1 2 - 3 444 107

View Full Product Details