Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-56
Entrez Gene ID
3565971
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F36G9.16
Chromosome Location
Chr.V: 15951432 - 15952862 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_001028718.1 NP_001023889.1 3 - 4 898 137

View Full Product Details