Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-20
Entrez Gene ID
185375
Gene Name
NRF domain-containing protein
Gene Aliases
CELE_F36G9.12
Chromosome Location
Chr.V: 15978714 - 15981779 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074606.5 NP_507007.3 6 - 7 1782 71

View Full Product Details