Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-35D1
Entrez Gene ID
184495
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F14H3.10
Chromosome Location
Chr.V: 16069238 - 16071318 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074643.2 NP_507044.1 1 - 2 203 78

View Full Product Details