Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
oac-15
Entrez Gene ID
184335
Gene Name
Acyl_transf_3 domain-containing protein
Gene Aliases
CELE_F11A5.8
Chromosome Location
Chr.V: 16206849 - 16209937 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074687.3 NP_507088.1 6 - 7 1439 97

View Full Product Details