Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
gst-22
Entrez Gene ID
180088
Gene Name
Glutathione S-Transferase
Gene Aliases
CELE_F21H7.1
Chromosome Location
Chr.V: 16231032 - 16233313 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074694.6 NP_507095.1 2 - 3 1157 70

View Full Product Details