Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cyp-37B1
Entrez Gene ID
180093
Gene Name
CYtochrome P450 family
Gene Aliases
CELE_F28G4.1
Chromosome Location
Chr.V: 16278116 - 16280616 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074708.7 NP_507109.1 2 - 3 635 95

View Full Product Details