Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srw-87
Entrez Gene ID
186467
Gene Name
G_PROTEIN_RECEP_F1_2 domain-containing protein
Gene Aliases
CELE_F57G8.4
Chromosome Location
Chr.V: 16336626 - 16338277 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074728.1 NP_507129.1 5 - 6 874 144

View Full Product Details