Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
sru-44
Entrez Gene ID
188299
Gene Name
Serpentine Receptor, class U
Gene Aliases
CELE_T08G3.8
Chromosome Location
Chr.V: 16466271 - 16467940 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074764.2 NP_507165.1 1 - 2 405 164

View Full Product Details