Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
srr-3
Entrez Gene ID
185351
Gene Name
Serpentine Receptor, class R
Gene Aliases
CELE_F36D3.3
Chromosome Location
Chr.V: 16506894 - 16508767 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074779.5 NP_507180.1 1 - 2 264 123

View Full Product Details