Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
clec-39
Entrez Gene ID
188899
Gene Name
C-type LECtin
Gene Aliases
CELE_T25E12.7
Chromosome Location
Chr.V: 16739268 - 16741588 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074835.4 NP_507236.3 5 - 6 570 93

View Full Product Details