Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cand-1
Entrez Gene ID
180125
Gene Name
Cullin-associated NEDD8-dissociated protein 1
Gene Aliases
CELE_Y102A5A.1
Chromosome Location
Chr.V: 16802872 - 16814957 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_074843.6 NP_507244.1 2 - 3 253 66

View Full Product Details