Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
str-77
Entrez Gene ID
189247
Gene Name
Seven TM Receptor
Gene Aliases
CELE_W06D12.4
Chromosome Location
Chr.V: 17718947 - 17721293 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075080.1 NP_507481.1 2 - 3 369 88

View Full Product Details