Assay Design

The assay probe spans an exon junction. This assay will detect gDNA.

Gene Transcripts

Gene Symbol
cutl-4
Entrez Gene ID
189246
Gene Name
ZP domain-containing protein
Gene Aliases
CELE_W06D12.1
Chromosome Location
Chr.V: 17733546 - 17739901 on Build WBcel235
Interrogated Sequence Translated Protein Exon Boundary Assay Location Amplicon Length
RefSeq NM_075083.6 NP_507484.3 6 - 7 1089 55

View Full Product Details